Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A 350619 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Activator of soluble guanylyl cyclase (sGC). Activates basal sGC and synergistically activates sGC in the presence of NO. Relaxes cavernosum smooth muscle in vitro and induces penile erections in rats. Induces a sustained increase in skin blood flow in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 425.41 |
Percent Purity | >99% |
Chemical Name or Material | A 350619 hydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction