Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 412997 dihydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
1347744-96-0
C19H25Cl2N3O
382.329
KIGNSSHMCNBHQX-UHFFFAOYSA-N
a-412997 dihydrochloride,a 412997 dihydrochloride,2-4-pyridin-2-yl piperidin-1-yl-n-m-tolyl acetamide dihydrochloride,n-3-methylphenyl-4-2-pyridinyl-1-piperidineacetamide dihydrochloride,n-3-methylphenyl-2-4-pyridin-2-yl piperidin-1-yl acetamide dihydrochloride
90488953
N-(3-methylphenyl)-2-(4-pyridin-2-ylpiperidin-1-yl)acetamide;dihydrochloride
CC1=CC(=CC=C1)NC(=O)CN2CCC(CC2)C3=CC=CC=N3.Cl.Cl
This item is not returnable. View return policy
1347744-96-0
C19H25Cl2N3O
382.329
KIGNSSHMCNBHQX-UHFFFAOYSA-N
a-412997 dihydrochloride,a 412997 dihydrochloride,2-4-pyridin-2-yl piperidin-1-yl-n-m-tolyl acetamide dihydrochloride,n-3-methylphenyl-4-2-pyridinyl-1-piperidineacetamide dihydrochloride,n-3-methylphenyl-2-4-pyridin-2-yl piperidin-1-yl acetamide dihydrochloride
90488953
N-(3-methylphenyl)-2-(4-pyridin-2-ylpiperidin-1-yl)acetamide;dihydrochloride
CC1=CC(=CC=C1)NC(=O)CN2CCC(CC2)C3=CC=CC=N3.Cl.Cl

Selective D4 agonist

Selective agonist for the dopamine D4 receptor (Ki values are 7.9 and 12.1 nM for human D4 and rat D4 receptors). Displays no affinity (<1000 nM) for other dopamine receptors. Shows efficacy in rat models of ADHD and short-term memory.
Quantity 10mg
Formula Weight 382.33
Percent Purity >98%
Chemical Name or Material A 412997 dihydrochloride

Recommended Storage : Desiccate at Room Temperature