Learn More
A 412997 dihydrochloride, Tocris™
Selective D4 agonist
Manufacturer:  R&D Systems 4552/50
Quantity 50mg
 View more versions of this product
Catalog No. 45-525-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
382.33
A 412997 dihydrochloride
C19H23N3O·2HCl
N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
Specifications
50mg
>98%
382.33
A 412997 dihydrochloride
1347744-96-0
C19H23N3O·2HCl
N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
Safety and Handling
Recommended Storage : Desiccate at Room Temperature