Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A-7 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
79127-24-5
C20H30Cl2N2O2S
433.432
XDJCAQBTSCRBHS-UHFFFAOYSA-N
a-7 hydrochloride, n-10-aminodecyl-5-chloro-1-naphthalenesulfonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide hydrochloride, 1-10-aminodec-1-yl sulphamoyl-5-chloronaphthalene, n-10-aminodecyl-5-chloro-1-naphthalenesulphonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide-hydrogen chloride 1/1
44119113
N-(10-aminodecyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride
C1=CC2=C(C=CC=C2Cl)C(=C1)S(=O)(=O)NCCCCCCCCCCN.Cl
This item is not returnable. View return policy
79127-24-5
C20H30Cl2N2O2S
433.432
XDJCAQBTSCRBHS-UHFFFAOYSA-N
a-7 hydrochloride, n-10-aminodecyl-5-chloro-1-naphthalenesulfonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide hydrochloride, 1-10-aminodec-1-yl sulphamoyl-5-chloronaphthalene, n-10-aminodecyl-5-chloro-1-naphthalenesulphonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide-hydrogen chloride 1/1
44119113
N-(10-aminodecyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride
C1=CC2=C(C=CC=C2Cl)C(=C1)S(=O)(=O)NCCCCCCCCCCN.Cl

Calmodulin antagonist

Potent calmodulin antagonist (inhibits calmodulin-activated PDE activity with an IC50 of 3 μM).
Quantity 10 mg
Formula Weight 433.44
Percent Purity >98%
Chemical Name or Material A-7 hydrochloride

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit