Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ A-803467, 98%

Click to view available options
Quantity:
5mg
10mg
50mg
944261-79-4
C19H16ClNO4
357.79
MFCD10574689
VHKBTPQDHDSBSP-UHFFFAOYSA-N
5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide,d0ff6s,2-furancarboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl,5-4-chlorophenyl-n-3,5-dimethoxyphenyl,5-4-chlorophenyl-furan-2-carboxylic acid 3,5,a hplc,5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furamide,2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl
16038374
5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
COC1=CC(=CC(=C1)NC(=O)C2=CC=C(O2)C3=CC=C(C=C3)Cl)OC
944261-79-4
C19H16ClNO4
357.79
MFCD10574689
VHKBTPQDHDSBSP-UHFFFAOYSA-N
5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide,d0ff6s,2-furancarboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl,5-4-chlorophenyl-n-3,5-dimethoxyphenyl,5-4-chlorophenyl-furan-2-carboxylic acid 3,5,a hplc,5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furamide,2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl
16038374
5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide
COC1=CC(=CC(=C1)NC(=O)C2=CC=C(O2)C3=CC=C(C=C3)Cl)OC
Quantity 10mg
Melting Point 128°C to 130°C
Color White
Formula Weight 357.79
Physical Form Solid
Percent Purity 98%
Chemical Name or Material A-803467

storageNote1 : -30° to -10°C