A 80426 mesylate, Tocris

High affinity α2 antagonist; Also 5-HT uptake inhibitor

Manufacturer: R&D Systems 2341/50

Quantity 50mg

 View more versions of this product

Catalog No. 23-415-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C23H27NO2·CH3SO3H
    Formula Weight 445.57
    Synonym N-Methyl-N-[[(1R)-1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl]methyl]-6-benzofuranethanamine mesylate
    Chemical Name or Material A 80426 mesylate

    Specifications

    Formula Weight 445.57
    Quantity 50mg
    Synonym N-Methyl-N-[[(1R)-1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl]methyl]-6-benzofuranethanamine mesylate
    Assay Percent Range >98%
    Molecular Formula C23H27NO2·CH3SO3H
    CAS 152148-64-6
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material A 80426 mesylate

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit