Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ A 939572
Potent SCD-1 inhibitor
$250.56 - $1066.04
Specifications
Chemical Name or Material | 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide |
---|---|
CAS | 1032229-3-6 |
Target | Compounds for Stem Cell Differentiation |
Molecular Formula | C20H22ClN3O3 |
Formula Weight | 387.86 |
Description
Potent stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 0.4 nM). Selective for SCD1 over a range of kinases and hERG channels. Inhibits proliferation of squamous cell carcinoma FaDu cells in vitro. Also induces cell death of undifferentiated human embryonic stem cells (ESCs). Orally bioavailable.Specifications
4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide | |
Compounds for Stem Cell Differentiation | |
387.86 |
1032229-3-6 | |
C20H22ClN3O3 | |
>99% |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title