Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 943931 dihydrochloride, Tocris Bioscience™

Catalog Number 375350
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1227675-50-4
C17H23Cl2N5
368.306
ISYUFRNZHUOGLA-CURYUGHLSA-N
a 943931 dihydrochloride, 4-3r-3-amino-pyrrolidin-1-yl-6,7-dihydro-5h-benzo 6,7 cyclohepta 1,2-d pyrimidin-2-ylamine
56972240
4-[(3R)-3-aminopyrrolidin-1-yl]-6,7-dihydro-5H-benzo[1,2]cyclohepta[3,4-b]pyrimidin-2-amine;dihydrochloride
C1CC2=CC=CC=C2C3=C(C1)C(=NC(=N3)N)N4CCC(C4)N.Cl.Cl
This item is not returnable. View return policy
1227675-50-4
C17H23Cl2N5
368.306
ISYUFRNZHUOGLA-CURYUGHLSA-N
a 943931 dihydrochloride, 4-3r-3-amino-pyrrolidin-1-yl-6,7-dihydro-5h-benzo 6,7 cyclohepta 1,2-d pyrimidin-2-ylamine
56972240
4-[(3R)-3-aminopyrrolidin-1-yl]-6,7-dihydro-5H-benzo[1,2]cyclohepta[3,4-b]pyrimidin-2-amine;dihydrochloride
C1CC2=CC=CC=C2C3=C(C1)C(=NC(=N3)N)N4CCC(C4)N.Cl.Cl

Potent and selective H4 antagonist

Potent and selective histamine H4 receptor antagonist (pKi values are 7.15 and 8.12 at human and rat receptors respectively). Blocks inflammation in a peritonitis mouse model and displays efficacy in inflammatory pain and neuropathic pain models.
Quantity 50 mg
Formula Weight 368.31
Percent Purity >98%
Chemical Name or Material A 943931 dihydrochloride
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit