Learn More
Tocris Bioscience™ A 971432 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective S1P5 agonist
$144.80 - $606.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-661-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5766/10
Each for $144.80
Add to cart
57-665-0 View Documents Tocris Bioscience™
5766/50
Each for $606.00
Add to cart
Description
Description
Potent and selective S1P5 agonist (EC50 values are 4.1 and 5.7 nM in cAMP and GTPγS assays, respectively). Exhibits 60-fold selectivity over S1P1, and >1,600-fold selectivity over S1P2-4, and a panel of 129 kinases and 79 receptors. Improves blood-brain barrier integrity in vitro and reverses age-related cognitive decline in vivo. Orally available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.