Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A.aeolicusBPL/BioID2 Mouse anti-Bacteria, Janelia Fluor 549, Clone: SS QD1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259940JF549
Description
A. aeolicus BPL/BioID2 Monoclonal specifically detects A. aeolicus BPL/BioID2 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
A.aeolicusBPL/BioID2 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GST fused to A. aeolicus BPL (Biotin Protein Ligase) R40G (BioID2) | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
SS QD1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
Epitope Tags | |
This antibody is reactive with A. aeolicus and has not been tested with other species. The birA Entrez Gene number is NP_213397. UniProt number is O66837. | |
Store at 4°C in the dark. | |
IgG2 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title