Learn More
A.aeolicusBPL/BioID2 Mouse anti-Bacteria, Janelia Fluor 646, Clone: SS QD1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259940JF646
Catalog No. NBP59940646
Add to cart
Description
Description
A.aeolicusBPL/BioID2 Monoclonal antibody specifically detects A.aeolicusBPL/BioID2 in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
A.aeolicusBPL/BioID2
SS QD1
Immunocytochemistry/Immunofluorescence
Purified
GST fused to A. aeolicus BPL (Biotin Protein Ligase) R40G (BioID2)
Protein A purified
Store at 4°C in the dark.
Monoclonal
Bacteria
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2 Kappa
0.1mL
Primary
This antibody is reactive with A. aeolicus and has not been tested with other species. The birA Entrez Gene number is NP_213397. UniProt number is O66837.