Learn More
Bachem Suc-Ala-Phe-Lys-AMC, 50mg, 73207-91-7, MFCD00151397, Acetate salt DFS Item
Manufacturer:  Bachem 4004022.0050
A highly sensitive fluorogenic substrate for plasmin. Suc-AFK-AMC was also used for assaying gingipain K.
Catalog No. 50-194-6938