Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A-RAF Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270201JF549

Catalog No. N270201J549


Add to cart

Description

Description

A-RAF Monoclonal antibody specifically detects A-RAF in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

A-RAF
OTI2G9
Immunohistochemistry
Affinity Purified, Conjugated
ARAF
Recombinant protein fragment corresponding to amino acids 145-365 of human A-RAF (NP_001645) produced in E.coli.
Immunogen affinity purified
Protein Kinase, Signal Transduction
Primary
369.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ARAF1A-RAF, EC 2.7.11, EC 2.7.11.1, Oncogene ARAF1, PKS, PKS2A-Raf proto-oncogene serine/threonine-protein kinase, Proto-oncogene A-Raf, Proto-oncogene A-Raf-1, Proto-oncogene Pks, RAFA1, Ras-binding protein DA-Raf, serine/threonine-protein kinase A-Raf, v-raf murine sarcoma 3611 viral oncogene homolog, v-raf murine sarcoma 3611 viral oncogene homolog 1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit