Learn More
Tocris Bioscience™ a2-AdrenoceptorTocris Bioscience™et™
Selection of 5 α2-adrenoceptor ligands (Cat. Nos. 1030, 0425, 0928, 1133 and 0891)
Supplier:  Tocris Bioscience™ 1872/1 SET
Catalog No. 18-721-R
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Contains the listed products either as lyophilised solids or pre-dissolved in DMSO to a stock concentration of 10 mM in 0.5 ml. The products can be used to investigate the pharmacology of α2-adrenoceptors.
Specifications
Specifications
-20°C
1 Set
α2-Adrenoceptor Tocriset™
α2-Adrenoceptor Tocriset™