Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A26C2/A26C3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550094

Catalog No. PA550094


Add to cart

Description

Description

The antibody detects endogenous levels of total A26C2 + A26C3 protein.

Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications

Specifications

A26C2/A26C3
Polyclonal
Unconjugated
POTEH
Liquid
POTEG, POTEH
Synthesized peptide derived from internal of human A26C2/3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6S5H5, Q6S545
A26C2, POTE14
Rabbit
IgG
100 μL
-20°C
Primary
23784, 404785
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit