Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A26C2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

$211.50 - $476.50

Specifications

Antigen A26C2
Concentration 0.5 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5122306
Encompass
View Documents
Thermo Scientific
PA5122306
20 μL
Each for $211.50
Add to Cart
 
PIPA5122305
Encompass
View Documents
Thermo Scientific
PA5122305
100 μL
Each for $476.50
Add to Cart
 
Description

Description

Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications

Specifications

A26C2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q6S5H5
404785
Ala146-Arg306 (Accession Q6S5H5)
Primary
-20°C
POTEG
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300
A26C2; ACTBL1; ANKRD26-like family C member 2; ANKRD26-like family C, member 2; cancer/testis antigen family 104, member 4; CT104.4; POTE ankyrin domain family member G; POTE ankyrin domain family, member G; POTE14; POTE-14; POTE14alpha; POTE22; POTEG; Prostate, ovary, testis-expressed protein on chromosome 14; protein expressed in prostate, ovary, testis, and placenta 14
POTEG
IgG
Antigen affinity chromatography
Antibody
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit