Learn More
Selleck Chemical LLCSupplier Diversity Partner A2AR antagonist 1 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85755MG
A2AR antagonist 1 is a potent A2AR (Adenosine A2A Receptor) antagonist with Ki values of 4 nM and 264 nM for A2AR and A1R, respectively.
Catalog No. 50-194-8416