Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A2M Goat anti-Rat, Polyclonal , Abnova™
Goat polyclonal antibody raised against rat A2m.
Supplier: Abnova Corporation PAB29697
Specifications
A2M | |
Polyclonal | |
Goat polyclonal antibody raised against rat A2m. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
A2m | |
Native purified rat A2m. | |
100 μg | |
Primary | |
Rat | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
A2MAC1/A2m1/A2maa/MGC114358/Mam/RATA2MAC1 | |
Goat | |
Antigen Affinity Purification | |
RUO | |
24153 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction