Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A33/GPA33 Antibody (2963D), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB11365F
Description
A33/GPA33 Monoclonal antibody specifically detects A33/GPA33 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
A33/GPA33 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
2963D | |
Western Blot, Flow Cytometry | |
A33, cell surface A33 antigen, Glycoprotein A33, glycoprotein A33 (transmembrane), MGC129986, MGC129987, transmembrane glycoprotein A33 | |
E. coli-derived recombinant human A33, Ile22-Val235, Accession # Q99795 | |
0.1 mL | |
Cancer | |
10223 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction