Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A33 Recombinant Rabbit Monoclonal Antibody (022), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$221.00 - $430.00
Specifications
Antigen | A33 |
---|---|
Clone | 022 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543102
![]() |
Thermo Scientific
MA543102 |
100 μL |
Each for $430.00
|
|
PIMA543445
![]() |
Thermo Scientific
MA543445 |
50 μL |
Each for $221.00
|
|
Description
Specificity: Human GPA33
A33 Recombinant Monoclonal antibody specifically detects A33 in Human samples. It is validated for ELISASpecifications
A33 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33 | |
Gpa33 | |
IgG | |
Protein A | |
Antibody |
022 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
Q99795 | |
10223 | |
Recombinant Human GPA33 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Gpa33 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title