Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A37, Tocris Bioscience™

ALDH1A1 inhibitor

$165.30 - $693.50

Chemical Identifiers

CAS 896795-60-1
Molecular Formula C21H25N3O3S2
Molecular Weight (g/mol) 431.569
InChI Key SKDRHRAYBYQVNU-UHFFFAOYSA-N
Synonym CM 037
PubChem CID 5049161
IUPAC Name ethyl 2-[[4-oxo-3-(3-pyrrolidin-1-ylpropyl)-[1]benzothiolo[3,2-d]pyrimidin-2-yl]sulfanyl]acetate
SMILES CCOC(=O)CSC1=NC2=C(C(=O)N1CCCN3CCCC3)SC4=CC=CC=C42
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-021-0 View Documents Tocris Bioscience
5802/10
10mg Each for $165.30
Add to cart
 
58-025-0 View Documents Tocris Bioscience
5802/50
50mg Each for $693.50
Add to cart
 
Description

Description

ALDH1A1 inhibitor (IC50 = 4.6 μM; Ki = 300 nM). Exhibits selectivity for ALDH1A1 over eight other ALDH isoforms. Disrupts spheroid formation and reduces cell viability of ovarian cancer cells in vitro.
Specifications

Chemical Identifiers

896795-60-1
431.569
CM 037
ethyl 2-[[4-oxo-3-(3-pyrrolidin-1-ylpropyl)-[1]benzothiolo[3,2-d]pyrimidin-2-yl]sulfanyl]acetate
C21H25N3O3S2
SKDRHRAYBYQVNU-UHFFFAOYSA-N
5049161
CCOC(=O)CSC1=NC2=C(C(=O)N1CCCN3CCCC3)SC4=CC=CC=C42

Specifications

Ethyl 2-[[3,4-dihydro-4-oxo-3-[3-(1-pyrrolidinyl)propyl][1]benzothieno[3,2-d]pyrimidin-2-yl]thio]acetate
>97%
CM 037
Solid
896795-60-1
C21H25N3O3S2
431.57
Safety and Handling

Safety and Handling

Recommended Storage : Soluble to 100mM in DMSO and to 100mM in ethanol

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit