Learn More
BiolegendSupplier Diversity Partner Alexa Fluor 488 anti-IRF4  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 646406
Alexa Fluor 488 anti-IRF4 [IRF4.3E4]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 ug
Catalog No. 50-712-555