Learn More
ATCC A549 Human Lung Epithelial Cell
Manufacturer:  ATCC CRMCCL185
Encompass
For use in testing and calibration in ISO 17025 accredited laboratories, to challenge assay performance, validate or compare test methods, and to establish sensitivity, linearity and specificity during assay validation or implementation. This cell line may be used as a transfection host. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. NC0964428