Learn More
BiolegendSupplier Diversity Partner Alexa Fluor 594 anti-mouse CD3  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 100240
Alexa Fluor 594 anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: IHC-F, FC, 3-D IHC; Size: 100 ug
Catalog No. 50-711-801