Learn More
BiolegendSupplier Diversity Partner Alexa Fluor 594 anti-mouse/human CD324 (E-Cadherin)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 147306
Alexa Fluor 594 anti-mouse/human CD324 (E-Cadherin) [DECMA-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Dog, Swine; Apps: ICC, IHC-P, IHC-F; Size: 100 ug
Catalog No. 50-711-919