Learn More
BiolegendSupplier Diversity Partner Alexa Fluor 594 Goat anti-mouse IgG (minimal x-reactivity)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 405326
Alexa Fluor 594 Goat anti-mouse IgG (minimal x-reactivity) [Poly4053]; Isotype: Goat Polyclonal IgG; Reactivity: Mouse; Apps: ICC, IHC-F, IHC-P; Size: 100 ug
Catalog No. 50-712-234