Learn More
BiolegendSupplier Diversity Partner Alexa Fluor 700 anti-mouse IgD  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 405729
Alexa Fluor 700 anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-712-237