Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AA 29504, Tocris™

Positive allosteric modulator of GABAA receptors

Manufacturer:  R&D Systems 3972/10

Quantity 10mg

 View more versions of this product

Catalog No. 39-7210


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>98%
327.42
AA 29504
C19H25N3O2
N1-Ethoxycarbonyl-N4-[(2,4,6-trimethylpheny)lmethyl]-1,2,4-triaminobenzene

Specifications

10mg
>98%
327.42
AA 29504
945828-50-2
C19H25N3O2
N1-Ethoxycarbonyl-N4-[(2,4,6-trimethylpheny)lmethyl]-1,2,4-triaminobenzene

Safety and Handling

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit