Learn More
Abnova™ AACS 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00065985T01
Catalog No. 89-932-048
Add to cart
Description
Description
Specifications
Specifications
293T
BC041000.1
pCMV-AACS full-length
100 μL
Human
AACS 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 45.6kDa
Transient overexpression cell lysate was tested with anti-AACS antibody (Mfr. No. H00065985-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot