Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AACS Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549422

Catalog No. PA549422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Activates acetoacetate to acetoacetyl-CoA. May be involved in utilizing ketone body for the fatty acid-synthesis during adipose tissue development.
Specifications

Specifications

AACS
Polyclonal
PBS with 0.09% sodium azide
Q86V21, Q9JMI1
Acetoacetyl-CoA synthetase, Acyl-CoA synthetase family member 1, Protein sur-5 homolog, AACS, ACSF1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65984, 65985
Western Blot
Unconjugated
AACS
Liquid
AACS
KLH conjugated synthetic peptide between 539-573 amino acids from the C-terminal region of human AACS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit