Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AAQ Chloride

$195.00 - $819.00

Specifications

Formula Weight Observed MW: 443.97
Chemical Name or Material N,N,N-Triethyl-2-oxo-2-[[4-[2-[4-[(1-oxo-2-propen-1-yl)amino]phenyl]diazenyl]phenylamino]ethanaminium chloride
Molecular Formula C23H30ClN5O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
546210 View Documents Tocris Bioscience™
5462/10
10mg Each for $195.00
Each for $195.00
Add to cart
 
546250 View Documents Tocris Bioscience™
5462/50
50mg Each for $819.00
Add to cart
 
Description

Description

Photoswitchable Kv channel blocker (IC50 values are 2 and 64 μM at 500 nm and 380 nm respectively). Switches conformation from cis to trans at 500 nm and trans to cis at 380 nm. Exhibits minimal activity at Nav1.2 and L-type Ca2+ channels. Stimulates action potential firing of hippocampal neurons in vitro at 500 nm and restores visual responsiveness in blind mice at 380 nm.
Specifications

Specifications

Observed MW: 443.97
N,N,N-Triethyl-2-oxo-2-[[4-[2-[4-[(1-oxo-2-propen-1-yl)amino]phenyl]diazenyl]phenylamino]ethanaminium chloride
C23H30ClN5O2
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit