Sino Biological™ AARS Recombinant Mouse Protein, His tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mouse AARS (Met 1-Asn 968) fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 50384M08B5

 View more versions of this product

Catalog No. 50-384-M08B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 108.3kd
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Source Mouse
    Structural Form Full Length, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name Alanyl-Trna Synthetasep
    Gene Symbol AARS
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Insect
    Product Type AARS Recombinant Mouse Protein
    For Use With (Application) Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 234734

    Expressed from DNA sequence encoding mouse AARS (Met 1-Asn 968) fused to polyhistidine tag at C-terminus

    • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit