Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AASD-PPT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | AASD-PPT |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP154977
|
Novus Biologicals
NBP154977 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
AASD-PPT Polyclonal specifically detects AASD-PPT in Human samples. It is validated for Western Blot.Specifications
AASD-PPT | |
Polyclonal | |
Rabbit | |
Q9NRN7 | |
60496 | |
IgG | |
36 kDa |
Western Blot | |
Unconjugated | |
RUO | |
4'-phosphopantetheinyl transferase, AASD-PPTDKFZp566E2346, Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase, aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, CGI-80, EC 2.7.8, EC 2.7.8.-, L-aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase, LYS2, LYS5, LYS5 ortholog | |
AASDHPPT | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title