Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AATK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550481

Catalog No. PA550481


Add to cart

Description

Description

The antibody detects endogenous levels of total AATK protein.

The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line.
Specifications

Specifications

AATK
Polyclonal
Unconjugated
AATK
Liquid
AATK
Synthetic peptide corresponding to residues near the C terminal of human Apoptosis-associated tyrosine kinase
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q6ZMQ8
AATYK, KIAA0641, LMR1, LMTK1
Rabbit
IgG
100 μL
-20°C
Primary
9625
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit