Learn More
Cell Biolabs IncSupplier Diversity Partner AAV-5 HELPER FREE PKG SYSTEM  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK405
The AAV-5 Helper Free Expression System uses a pHelper plasmid to eliminate the need for a helper virus. Contains all packaging vectors plus an expression vector into which a gene plus its optimal promoter are cloned.
Catalog No. 50-673-811