Learn More
Cell Biolabs IncSupplier Diversity Partner AAV-6 PROMOTERLESS EXP SYSTEM  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK411SER6
The AAV-6 Helper Free Promoterless Expression System uses a pHelper plasmid to eliminate the need for a helper virus. Contains all packaging vectors plus an expression vector into which a gene plus its optimal promoter are cloned.
Catalog No. 50-673-829