Learn More
Cell Biolabs IncSupplier Diversity Partner AAV PURIFICATION KT 2 PREP  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK140T
ViraBind AAV Purification Kit (Trial Size) produces higher viral yield with higher purity than ultracentrifugation in a fraction of the time. Each prep can purify supernatant from two 10cm dishes or one 15cm dish. 2 preps/kit.
Catalog No. 50-673-755
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.