Learn More
Cell Biolabs IncSupplier Diversity Partner AAV PURIFICATN MEGA KT 2 PREP  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK141
ViraBind AAV Purification Mega Kit produces higher viral yield with higher purity than ultracentrifugation in a fraction of the time. Each prep can purify supernatant from up to ten 15cm dishes. 2 preps/kit.
Catalog No. 50-673-756