Learn More
Cell Biolabs IncSupplier Diversity Partner AAV QUANTITATION KIT TRL SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK145T
The QuickTiter AAV Quantitation Kit (Trial Size) provides an ultra-fast 45-60 minute procedure that measures physical titer via the viral nucleic acid content of unpurified AAV-2 or purified AAV of any serotype. 4 assays/kit.
Catalog No. 50-673-758