Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAV Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$206.00 - $436.00
Specifications
Antigen | VP1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB193575SS
|
Novus Biologicals
NB10093575SS |
0.025 mg |
Each for $206.00
|
|
NB10093575
|
Novus Biologicals
NB10093575 |
0.1 mg |
Each for $436.00
|
|
Description
VP1 Polyclonal specifically detects VP1 in Adeno-associated Virus (AAV) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VP1 | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Rabbit | |
Adeno-associated Virus (AAV), Virus | |
P03135 | |
17 | |
Synthetic peptide within residues 70-90 of major coat protein VP3 [AAV 2], residues 270-290 of capsid protein VP1 [AAV 6], and residues 270-290 of capsid AAV 3]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
AAV, AAV2, Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
VP1 | |
IgG | |
Affinity Purified | |
Adeno-Associated Virus (AAV) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title