Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AAV5 VP3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093577AF594
Description
AAV5 VP3 Polyclonal antibody specifically detects AAV5 VP3 in Nonspecies specific, Virus samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
AAV5 VP3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 594 | |
Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5. | |
0.1 mL | |
Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
All species, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction