Learn More
BioVision IncSupplier Diversity Partner EZLabel™ Antibody FITC Labeling Kit: Easy to use Kit to label upto 5 mg of Antibody with FITC for multiple applications such as ELISA, Western blot, Immunohistochemistry, Immunofluorescence and FACS analysis DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K8315
BioVision’s EZLabel Antibody-FITC Labeling Kit provides an easy way to label antibodies with Fluorescein Isothiocyanate (FITC) in a user-friendly spin column format. FITC is an ideal dye for fluorescent labeling of antibodies among the most commonly used fluorescent dyes for labeling antibodies. Each spin column provided in the kit can be used to label up to 1 mg of the target antibody. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of antibody per reaction. FITC-labeled antibody has an excitation and emission wavelengths at 494 nm and 520 nm respectively, and can be directly used for downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, and FACS analysis, etc. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9945