Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABAT Mouse anti-Human, Clone: UMAB180, liquid, UltraMAB™

Mouse Monoclonal Antibody
Supplier: OriGene UM800072

Description
ABAT (4-aminobutyrate aminotransferase) is responsible for catabolism of gamma-aminobutyric acid (GABA), an important, mostly inhibitory neurotransmitter in the central nervous system, into succinic semialdehyde. The active enzyme is a homodimer of 50-kD subunits complexed to pyridoxal-5-phosphate. The protein sequence is over 95% similar to the pig protein. GABA is estimated to be present in nearly one-third of human synapses. ABAT in liver and brain is controlled by 2 codomit alleles with a frequency in a Caucasian population of 0.56 and 0.44. The ABAT deficiency phenotype includes psychomotor retardation, hypotonia, hyperreflexia, lethargy, refractory seizures, and EEG abnormalities. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.Specifications
ABAT | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P80404 | |
ABAT | |
Human recombit protein fragment corresponding to amino acids 29-323 of human ABAT produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
UMAB180 | |
Unconjugated | |
ABAT | |
GABA-AT, GABAT, NPD009 | |
Mouse | |
Affinity Chromatography | |
RUO | |
18 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title