Abbott i-STAT™ 1 Wireless Handheld Point-of-Care Testing System
Integrated wireless capability allows you to test and transmit from the bedside
Manufacturer: Abbott Laboratories 03P7501
 View more versions of this product
Catalog No. 03P75-01A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Alert
For in vitro diagnostic use only
Required Accessories : CDS or DE with third-party Data Manager
    Specifications
    Certifications/Compliance FCC ID, IEC 61010-1: 2nd. Ed., U.S. 21 CFR 1040.10 and 1040.11, FCC, EN 60825-1:1994 + A1:2002 + A2:2001, IEC 60825-1:1993 + A1:1997 + A2:2001
    Compatibility 802.11b/802.11g wireless networks
    For Use With (Application) For testing blood gases, electrolytes, chemistries, coagulation, hematology, glucose and cardiac markers (cTnI)
    For Use With (Equipment) For LAN filter cable
    Includes i-STAT 1 wireless Handheld unit, battery carrier with two 9V disposable batteries, user guide
    Quantity Each
    Product Type i-STAT 1 Wireless Handheld Analyzer
  • Description & Specifications
Related Products
Product Content Feedback