Learn More
Abbott Abbott Point of Care i-STAT™ Tricontrols Materials
Manufacturer:  Abbott 05P7301
 View more versions of this product
Catalog No. 05-P73-01A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Healthcare
3
2° to 8°C
Liquid QC and calibration verification, measures hematocrit, Blood gases, electrolytes, chemistries
TriControls Control Level 3
Na: 150mM/L, K: 6.3mM/L
1.7mL