Abbott Point of Care i-STAT™ Tricontrols Materials

Manufacturer: Abbott Laboratories 05P7001

 View more versions of this product

Catalog No. 05-P70-01A

  Check Availability
Add to Cart

    Specifications

    Product Type Tricontrols calibration verification set
    For Use With (Application) Liquid QC and calibration verification, measures hematocrit, Blood gases, electrolytes, chemistries
    Storage Requirements 2° to 8°C
    Quantity 1.7mL,1.7mL
    Level 1 to 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit