Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCA7 Monoclonal Antibody (7A1-144), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA524784
Description
ABCA7 Monoclonal antibody specifically detects ABCA7 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ABCA7 | |
Monoclonal | |
1.3 mg/mL | |
tris glycine with 0.15M NaCl and 0.05% sodium azide | |
Q91V24 | |
Abca7 | |
Mouse ABCA7 stably-transfected HeLa cells. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
7A1-144 | |
Unconjugated | |
Abca7 | |
ABC 1 antibody; ABC Transporter 1; ABC1; ABC1 antibody; Abc51; Abca7; ABCA-SSN; ABCX; AD9; ATP binding cassette subfamily A member 7; ATP-binding cassette sub-family A member 7; ATP-binding cassette sub-family A member 7; LOW QUALITY PROTEIN: ATP-binding cassette sub-family A member 7; ATP-binding cassette, subfamily A (ABC1), member 7; ATP-binding cassette, sub-family A (ABC1), member 7; ATP-binding cassette, sub-family A, member 7; autoantigen SS-N; CERP; CERP antibody; EC 2.2.1.1; EC 3.6.3; EC 3.6.3.41; FLJ40025; Macrophage ABC transporter; Phospholipid-transporting ATPase ABCA7; phospholipid-transporting ATPase ABCA7; ATP-binding cassette sub-family A member 7 | |
Rat | |
Protein G | |
RUO | |
27403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction