Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCB1, anti-Human, Clone: MRK16, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00101
Specifications
ABCB1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
ABC20/CD243/CLCS/GP170/MDR1/MGC163296/P-GP/PGY1 | |
Original antibody is raised against drug resistant K-562 cells. | |
RUO | |
5243 | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
MRK16 | |
Flow Cytometry (1 ug/mL) Immunofluorescence Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user. | |
ATP-binding cassette, sub-family B (MDR/TAP), member 1 | |
ABCB1 | |
200 μg | |
Primary | |
This antibody recognizes a discontinuous extracellular epitope on ABCB1. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction