Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCB10 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169066
Description
ABCB10 Polyclonal specifically detects ABCB10 in Mouse samples. It is validated for Western Blot.Specifications
ABCB10 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9JI39 | |
Abcb10 | |
Synthetic peptides corresponding to Abcb10 (ATP-binding cassette, sub-family B (MDR/TAP), member 10) The peptide sequence was selected from the N terminal of Abcb10. Peptide sequence NGIRVYLMQSSGQSIVNRLRTSLFSSILRQEVAFFDKTRTGELINRLSSD. | |
Affinity Purified | |
RUO | |
23456 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ABC transporter 10 protein, ATP-binding cassette, sub-family B (MDR/TAP), member 10, EC 3.6.3, EC 3.6.3.44, EST20237, M-ABC2ATP-binding cassette transporter 10, Mitochondrial ATP-binding cassette 2, MTABC2ATP-binding cassette sub-family B member 10, mitochondrial | |
Rabbit | |
77 kDa | |
100 μL | |
Primary | |
Goat: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title