Learn More
ABCB5 Monoclonal Antibody (5H3C6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA517026
Description
MA5-17026 targets ABCB5 in FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17026 immunogen is purified recombit fragment of human ABCB5 (amino acids: 481-674) expressed in E. Coli. MA5-17026 detects ABCB5 which has a predicted molecular weight of approximately 89.8kDa.
ABCB5 belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules
Specifications
ABCB5 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q2M3G0 | |
Mouse | |
Purified | |
RUO | |
340273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
5H3C6 | |
Unconjugated | |
ABCB5 | |
ABCB5 | |
Purified recombit fragment of human ABCB5 (amino acids: 481-674) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.